siRNA Details
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 24 | 2987 | ||
XM_017023023.1 | XP_016878512.1 | 24 | 3005 | |||
XM_017023024.1 | XP_016878513.1 | 6 | 618 | |||
GenBank mRNA | AB067511.1 | - | 21 | 5016 | ||
AK093802.1 | - | 4 | 364 | |||
AK126622.1 | - | 10 | 1626 | |||
AK131510.1 | - | 7 | 911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
