siRNA Details
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 22 | 2773 | ||
XM_017023023.1 | XP_016878512.1 | 22 | 2776 | |||
XM_017023024.1 | XP_016878513.1 | 4 | 448 | |||
GenBank mRNA | AB067511.1 | - | 19 | 4802 | ||
AK093802.1 | - | 2 | 150 | |||
AK126622.1 | - | 8 | 1412 | |||
AK131510.1 | - | 5 | 697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
