siRNA Details
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 8 | 2023 | ||
NM_001281988.1 | NP_001268917.1 | 7 | 601 | |||
NM_144569.5 | NP_653170.3 | 8 | 2023 | |||
XM_011542413.2 | XP_011540715.1 | 8 | 592 | |||
XM_011542414.2 | XP_011540716.1 | 7 | 787 | |||
XM_017002779.1 | XP_016858268.1 | 7 | 1932 | |||
XM_017002780.1 | XP_016858269.1 | 6 | 1890 | |||
XM_017002781.1 | XP_016858270.1 | 7 | 1932 | |||
XM_017002782.1 | XP_016858271.1 | 7 | 1161 | |||
GenBank mRNA | AK091016.1 | - | 7 | 572 | ||
AK095829.1 | - | 1 | 199 | |||
AK096192.1 | - | 1 | 55 | |||
AK131447.1 | - | 8 | 1952 | |||
BC007910.2 | - | 1 | 89 | |||
BC029069.1 | - | 1 | 39 | |||
BC114430.1 | - | 8 | 1949 | 40038407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
