siRNA Details
Gene Symbol: ZDHHC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039617.1 | NP_001034706.1 | 1 | 247 | ||
NR_135617.1 | 1 | 255 | ||||
NR_135618.1 | 1 | 255 | ||||
XM_005269279.2 | XP_005269336.1 | 1 | 255 | |||
XM_006713493.2 | XP_006713556.1 | 1 | 231 | |||
XM_006713494.2 | XP_006713557.1 | 1 | 231 | |||
XM_011512417.1 | XP_011510719.1 | 1 | 134 | |||
GenBank mRNA | AK122907.1 | - | 1 | 255 | ||
BC022078.2 | - | 1 | 247 | 4827355 | ||
BG721568.1 | - | 1 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
