siRNA Details
Gene Symbol: FAM170A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163991.1 | NP_001157463.1 | 2 | 988 | ||
NM_182761.3 | NP_877438.2 | 3 | 1129 | |||
GenBank mRNA | AF427126.1 | - | 3 | 933 | ||
AY692447.1 | - | 2 | 988 | |||
BC040622.1 | - | 3 | 1090 | |||
BC128243.1 | - | 3 | 1002 | 40114744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
