siRNA Details
Gene Symbol: FMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197964.4 | NP_932068.2 | 2 | 611 | ||
NR_073058.1 | 3 | 484 | ||||
NR_073059.1 | 2 | 390 | ||||
NR_073060.1 | 2 | 370 | ||||
GenBank mRNA | AA393224.1 | - | 2 | 326 | ||
AF161386.1 | - | 2 | 416 | |||
BC008467.1 | - | 2 | 414 | 4276817 | ||
BI117784.1 | - | 2 | 611 | |||
BU852633.1 | - | 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
