siRNA Details
Gene Symbol: C7orf55-LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244584.2 | NP_001231513.1 | 1 | 366 | ||
GenBank mRNA | AK297642.1 | - | 1 | 138 |
Gene Symbol: FMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197964.4 | NP_932068.2 | 1 | 366 | ||
GenBank mRNA | AF161386.1 | - | 1 | 172 | ||
AK093018.1 | - | 1 | 135 | |||
BC008467.1 | - | 1 | 169 | 4276817 | ||
BI117784.1 | - | 1 | 366 | |||
CB125801.1 | - | 1 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
