siRNA Details
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145914.2 | NP_666019.1 | 4 | 1447 | ||
XM_005250568.4 | XP_005250625.1 | 5 | 2589 | |||
XM_006716113.3 | XP_006716176.1 | 4 | 2428 | |||
XM_017012584.1 | XP_016868073.1 | 5 | 2553 | |||
XM_017012585.1 | XP_016868074.1 | 5 | 1368 | |||
GenBank mRNA | AF155100.1 | - | 3 | 1122 | ||
AK075324.1 | - | 6 | 1444 | |||
AK292267.1 | - | 4 | 1467 | |||
AK301871.1 | - | 6 | 1498 | |||
AK313884.1 | - | 4 | 1465 | |||
AL136865.1 | - | 3 | 1400 | |||
BC033129.1 | - | 1 | 572 | |||
BC047309.1 | - | 4 | 1440 | 5111890 | ||
CR749301.1 | - | 5 | 1331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
