siRNA Details
Gene Symbol: GREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191322.1 | NP_001178251.1 | 3 | 397 | ||
NM_001191323.1 | NP_001178252.1 | 3 | 484 | |||
NM_013372.6 | NP_037504.1 | 2 | 607 | |||
XM_005254301.2 | XP_005254358.1 | 2 | 671 | |||
XM_017022077.1 | XP_016877566.1 | 2 | 653 | |||
GenBank mRNA | AF045800.1 | - | 1 | 448 | ||
AF110137.2 | - | 2 | 510 | |||
AF154054.1 | - | 2 | 577 | |||
AK095741.1 | - | 2 | 451 | |||
AK095890.1 | - | 3 | 484 | |||
AK095972.1 | - | 3 | 397 | |||
AK312265.1 | - | 2 | 608 | |||
AY232290.1 | - | 2 | 583 | |||
BC069525.1 | - | 1 | 448 | 7262108 | ||
BC093778.1 | - | 2 | 470 | 7939623 | ||
BC101611.1 | - | 2 | 470 | 8069117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
