siRNA Details
Gene Symbol: NKX3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256339.1 | NP_001243268.1 | 3 | 316 | ||
NM_006167.3 | NP_006158.2 | 2 | 589 | |||
GenBank mRNA | AF247704.1 | - | 2 | 589 | ||
AF249669.1 | - | 3 | 419 | |||
AF249670.1 | - | 2 | 404 | |||
AF249671.1 | - | 3 | 344 | |||
AF249672.1 | - | 3 | 326 | |||
BC074863.2 | - | 2 | 573 | 30915465 | ||
BC074864.2 | - | 2 | 573 | 30915229 | ||
U80669.1 | - | 2 | 570 | |||
U91540.1 | - | 2 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
