siRNA Details
Gene Symbol: ARL6IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002251.2 | NP_001002251.2 | 3 | 974 | ||
NM_001002252.2 | NP_001002252.2 | 3 | 941 | |||
NM_001278378.1 | NP_001265307.1 | 3 | 974 | |||
NM_001278379.1 | NP_001265308.1 | 3 | 581 | |||
NM_001278380.1 | NP_001265309.1 | 3 | 1107 | |||
NM_016638.3 | NP_057722.3 | 3 | 941 | |||
NM_018694.3 | NP_061164.3 | 3 | 998 | |||
NR_103512.1 | 3 | 913 | ||||
GenBank mRNA | AB035384.1 | - | 3 | 306 | ||
AF267748.1 | - | 3 | 389 | |||
AK027285.1 | - | 2 | 643 | |||
AK315759.1 | - | 3 | 297 | |||
BC001958.1 | - | 3 | 275 | 3544177 | ||
BC015569.2 | - | 3 | 271 | 3609127 | ||
BC015909.1 | - | 3 | 998 | 4648478 | ||
BC094839.1 | - | 3 | 708 | 6339167 | ||
BQ643520.1 | - | 3 | 378 | |||
CD672894.1 | - | 3 | 324 | |||
DQ099385.1 | - | 2 | 277 | |||
EF036485.1 | - | 2 | 262 | |||
EU624490.1 | - | 3 | 941 | |||
EU624491.1 | - | 3 | 672 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144509, AB035383, AK133934, BC063748.
Gene Ontology Categories:
