siRNA Details
Gene Symbol: ZNF646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014699.3 | NP_055514.3 | 2 | 4206 | ||
XM_005255710.3 | XP_005255767.1 | 2 | 4985 | |||
XM_005255711.3 | XP_005255768.1 | 2 | 4944 | |||
XM_005255712.3 | XP_005255769.1 | 2 | 4985 | |||
XM_011545990.2 | XP_011544292.1 | 2 | 4211 | |||
GenBank mRNA | AB002294.1 | - | 1 | 4340 | ||
BC035589.1 | - | 2 | 4192 | 5532753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
