siRNA Details
Gene Symbol: CD320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165895.1 | NP_001159367.1 | 4 | 798 | ||
NM_016579.3 | NP_057663.1 | 5 | 924 | |||
GenBank mRNA | AF161254.1 | - | 5 | 923 | ||
AK058014.1 | - | 4 | 792 | |||
AK222623.1 | - | 5 | 908 | |||
AL136652.1 | - | 5 | 913 | |||
AL365453.1 | - | 2 | 354 | |||
AL365455.1 | - | 5 | 896 | |||
AY358420.1 | - | 5 | 890 | |||
BC000668.2 | - | 5 | 895 | 3347569 | ||
BC007083.1 | - | 5 | 878 | 4076237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
