siRNA Details
Gene Symbol: STEAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205315.1 | NP_001192244.1 | 3 | 461 | ||
NM_001205316.1 | NP_001192245.1 | 2 | 362 | |||
NM_024636.3 | NP_078912.2 | 2 | 362 | |||
GenBank mRNA | AF423422.1 | - | 3 | 344 | ||
AF423423.1 | - | 3 | 344 | |||
AK026806.1 | - | 2 | 341 | |||
AK307644.1 | - | 2 | 340 | |||
BC020600.1 | - | 2 | 337 | 4616389 | ||
DA977808.1 | - | 2 | 343 | |||
DB162772.1 | - | 3 | 442 | |||
DC333286.1 | - | 2 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
