siRNA Details
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024641.3 | NP_078917.2 | 2 | 205 | ||
XM_005267147.3 | XP_005267204.1 | 2 | 598 | |||
GenBank mRNA | AJ577574.1 | - | 1 | 70 | ||
AK291940.1 | - | 2 | 136 | |||
AY048774.1 | - | 2 | 160 | |||
AY048775.1 | - | 2 | 160 | |||
AY372528.1 | - | 1 | 25 | |||
BC137014.1 | - | 1 | 74 | 9021007 | ||
BC137016.1 | - | 1 | 74 | 9021009 | ||
BC146671.1 | - | 2 | 140 | 40148201 | ||
BC150199.1 | - | 2 | 140 | 40148811 | ||
BX537398.1 | - | 2 | 136 | |||
BX640869.1 | - | 2 | 166 | |||
CR936827.1 | - | 2 | 140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
