siRNA Details
Gene Symbol: ZNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006526.2 | NP_006517.1 | 3 | 3210 | ||
XM_005260545.3 | XP_005260602.1 | 4 | 3440 | |||
XM_006723875.2 | XP_006723938.1 | 6 | 3621 | |||
XM_011529036.1 | XP_011527338.1 | 6 | 3583 | |||
XM_017028059.1 | XP_016883548.1 | 4 | 3365 | |||
GenBank mRNA | AF041259.1 | - | 3 | 3210 | ||
AK290350.1 | - | 4 | 3437 | |||
BC113427.1 | - | 3 | 3038 | 8322479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
