siRNA Details
Gene Symbol: ZNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006526.2 | NP_006517.1 | 5 | 3508 | ||
XM_005260545.3 | XP_005260602.1 | 5 | 3605 | |||
XM_006723875.2 | XP_006723938.1 | 7 | 3786 | |||
XM_011529036.1 | XP_011527338.1 | 7 | 3748 | |||
XM_017028059.1 | XP_016883548.1 | 6 | 3663 | |||
GenBank mRNA | AF041259.1 | - | 5 | 3508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
