siRNA Details
Gene Symbol: FAM117B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173511.3 | NP_775782.2 | 3 | 845 | ||
GenBank mRNA | AB053315.1 | - | 3 | 542 | ||
AK096090.1 | - | 3 | 429 | |||
BC065010.1 | - | 2 | 251 | |||
BC106906.2 | - | 2 | 194 | 40031780 | ||
BC106907.2 | - | 2 | 190 | 40031781 | ||
CN397358.1 | - | 4 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
