siRNA Details
Gene Symbol: ZNF30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099437.1 | NP_001092907.1 | 5 | 2224 | ||
NM_001099438.1 | NP_001092908.1 | 5 | 2290 | |||
NM_194325.2 | NP_919306.2 | 5 | 2221 | |||
NR_024018.1 | 6 | 2301 | ||||
XM_011527443.2 | XP_011525745.1 | 5 | 2340 | |||
XM_011527444.1 | XP_011525746.1 | 5 | 1929 | |||
XM_017027423.1 | XP_016882912.1 | 4 | 2549 | |||
XM_017027424.1 | XP_016882913.1 | 5 | 1975 | |||
XM_017027425.1 | XP_016882914.1 | 5 | 2280 | |||
XM_017027426.1 | XP_016882915.1 | 5 | 2736 | |||
XM_017027427.1 | XP_016882916.1 | 6 | 2316 | |||
XM_017027428.1 | XP_016882917.1 | 6 | 2320 | |||
XM_017027429.1 | XP_016882918.1 | 6 | 2347 | |||
GenBank mRNA | AK000569.1 | - | 1 | 1374 | ||
AK295515.1 | - | 5 | 2080 | |||
BC142996.1 | - | 5 | 2209 | 8860451 | ||
BX640666.1 | - | 6 | 2224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
