siRNA Details
Gene Symbol: TAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153365.2 | NP_699196.2 | 4 | 616 | ||
XM_005248139.2 | XP_005248196.1 | 3 | 430 | |||
XM_005248140.3 | XP_005248197.1 | 3 | 380 | |||
XM_011513812.2 | XP_011512114.1 | 4 | 614 | |||
XM_011513815.2 | XP_011512117.1 | 3 | 1715 | |||
XM_011513816.2 | XP_011512118.1 | 3 | 424 | |||
XM_017007876.1 | XP_016863365.1 | 5 | 506 | |||
GenBank mRNA | AF189251.1 | - | 2 | 274 | ||
AK074494.1 | - | 4 | 549 | |||
AK296104.1 | - | 3 | 418 | |||
AK299163.1 | - | 1 | 47 | |||
BC066899.1 | - | 4 | 554 | 4825556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
