siRNA Details
Gene Symbol: DOCK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144658.3 | NP_653259.3 | 39 | 4224 | ||
XM_005262368.3 | XP_005262425.1 | 40 | 4508 | |||
XM_005262369.4 | XP_005262426.1 | 40 | 4202 | |||
XM_011531276.2 | XP_011529578.1 | 40 | 4143 | |||
XM_011531278.2 | XP_011529580.1 | 27 | 2629 | |||
GenBank mRNA | AK056684.1 | - | 9 | 738 | ||
AK124735.1 | - | 34 | 3647 | |||
AK125641.1 | - | 18 | 1736 | |||
AY692226.1 | - | 39 | 4224 | |||
BC047713.1 | - | 5 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
