siRNA Details
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129888.1 | NP_001123360.1 | 2 | 339 | ||
NM_032687.3 | NP_116076.1 | 2 | 375 | |||
NM_138496.1 | NP_612505.1 | 1 | 200 | |||
XM_006716560.3 | XP_006716623.1 | 2 | 592 | |||
XM_006716561.3 | XP_006716624.1 | 2 | 544 | |||
XM_017013463.1 | XP_016868952.1 | 2 | 600 | |||
XM_017013464.1 | XP_016868953.1 | 2 | 369 | |||
XM_017013465.1 | XP_016868954.1 | 2 | 487 | |||
XM_017013466.1 | XP_016868955.1 | 2 | 369 | |||
GenBank mRNA | AK094514.1 | - | 1 | 200 | ||
BC005073.1 | - | 2 | 375 | 3533499 | ||
BC056412.1 | - | 1 | 653 | |||
BC065484.1 | - | 2 | 339 | 4638402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
