siRNA Details
Gene Symbol: ZNF672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024836.2 | NP_079112.1 | 4 | 1074 | ||
XM_005270336.1 | XP_005270393.1 | 4 | 919 | |||
GenBank mRNA | AK025954.1 | - | 4 | 921 | ||
AK027476.1 | - | 1 | 99 | |||
AK027679.1 | - | 2 | 468 | |||
BC008872.1 | - | 4 | 729 | |||
BC029310.1 | - | 4 | 972 | |||
BC035140.1 | - | 1 | 1454 | 5264126 | ||
BC050348.1 | - | 1 | 1450 | 5260682 | ||
BC068506.1 | - | 4 | 772 | 4824147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
