siRNA Details
Gene Symbol: C17orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033046.3 | NP_001028218.1 | 7 | 786 | ||
NM_001100407.2 | NP_001093877.1 | 8 | 836 | |||
NM_001100408.2 | NP_001093878.1 | 6 | 744 | |||
NM_001193653.1 | NP_001180582.1 | 8 | 1056 | |||
NM_001193654.1 | NP_001180583.1 | 7 | 1006 | |||
NM_001193655.1 | NP_001180584.1 | 7 | 950 | |||
NM_001193657.1 | NP_001180586.1 | 8 | 1003 | |||
NR_036514.1 | 6 | 712 | ||||
NR_036516.1 | 6 | 663 | ||||
NR_036517.1 | 5 | 621 | ||||
NR_036518.1 | 4 | 547 | ||||
XM_006722293.2 | XP_006722356.1 | 6 | 1511 | |||
XM_011523606.2 | XP_011521908.1 | 6 | 1691 | |||
XM_017025072.1 | XP_016880561.1 | 6 | 1691 | |||
XM_017025073.1 | XP_016880562.1 | 5 | 1649 | |||
XM_017025074.1 | XP_016880563.1 | 7 | 784 | |||
XM_017025075.1 | XP_016880564.1 | 7 | 1553 | |||
XM_017025076.1 | XP_016880565.1 | 8 | 781 | |||
XM_017025077.1 | XP_016880566.1 | 7 | 945 | |||
XM_017025078.1 | XP_016880567.1 | 6 | 742 | |||
GenBank mRNA | AF269290.1 | - | 2 | 913 | ||
AK027743.1 | - | 4 | 487 | |||
AK056342.1 | - | 6 | 619 | |||
AK074950.1 | - | 8 | 933 | |||
AK090484.1 | - | 6 | 2857 | |||
AK092865.1 | - | 4 | 1845 | |||
AK225389.1 | - | 7 | 692 | |||
BC003595.1 | - | 7 | 670 | 2822930 | ||
BC004171.2 | - | 7 | 670 | 2822930 | ||
BM147030.1 | - | 6 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
