siRNA Details
Gene Symbol: OR51I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005288.2 | NP_001005288.1 | 1 | 477 | ||
GenBank mRNA | BC151148.1 | - | 1 | 477 | 9021023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
