siRNA Details
Gene Symbol: SAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168682.1 | NP_001162153.1 | 4, 5 | 397 | ||
XM_005250096.3 | XP_005250153.1 | 4, 5 | 635 | |||
XM_006715822.3 | XP_006715885.1 | 4, 5 | 587 | |||
GenBank mRNA | BQ635892.1 | - | 4, 5 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
