siRNA Details
Gene Symbol: PARP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317895.1 | NP_001304824.1 | 7 | 2509 | ||
NM_032789.4 | NP_116178.2 | 8 | 2385 | |||
XM_011517336.2 | XP_011515638.1 | 8 | 2325 | |||
GenBank mRNA | AK027370.1 | - | 8 | 2342 | ||
AK075250.1 | - | 4 | 1072 | |||
AK124017.1 | - | 1 | 1043 | |||
AK128537.1 | - | 9 | 2200 | |||
AK129821.1 | - | 4 | 863 | |||
AK222914.1 | - | 8 | 2368 | |||
AK303317.1 | - | 7 | 2434 | |||
BC014229.2 | - | 1 | 369 | |||
BC019030.2 | - | 4 | 1205 | |||
BC020378.1 | - | 8 | 2332 | |||
BC030549.1 | - | 8 | 2359 | |||
BC132961.1 | - | 8 | 2336 | 40146983 | ||
BC136542.1 | - | 8 | 2336 | 9020532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
