siRNA Details
Gene Symbol: LUZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142546.1 | NP_001136018.1 | 3 | 3226 | ||
NM_033631.3 | NP_361013.3 | 4 | 3292 | |||
XM_011542090.2 | XP_011540392.1 | 5 | 3776 | |||
XM_011542091.2 | XP_011540393.1 | 4 | 3710 | |||
XM_017002252.1 | XP_016857741.1 | 6 | 3950 | |||
GenBank mRNA | AK074153.1 | - | 1 | 2887 | ||
BC033219.1 | - | 1 | 343 | |||
BC051733.1 | - | 2 | 3079 | 4838475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
