siRNA Details
Gene Symbol: SUPT6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320755.1 | NP_001307684.1 | 37 | 5253 | ||
NM_003170.4 | NP_003161.2 | 36 | 5151 | |||
XM_017024978.1 | XP_016880467.1 | 38 | 5312 | |||
XM_017024979.1 | XP_016880468.1 | 36 | 5389 | |||
XM_017024980.1 | XP_016880469.1 | 35 | 5083 | |||
GenBank mRNA | AF070532.1 | - | 5 | 600 | ||
AL833607.1 | - | 37 | 5254 | |||
BC003692.1 | - | 8 | 1143 | |||
BC003696.1 | - | 21 | 3026 | |||
BC033074.1 | - | 13 | 1679 | |||
BC073963.1 | - | 9 | 1450 | |||
BC136522.1 | - | 36 | 5063 | 9020512 | ||
BC136524.1 | - | 36 | 5063 | 9020514 | ||
BC150268.1 | - | 36 | 5065 | 9007230 | ||
D79984.1 | - | 36 | 5065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
