siRNA Details
Gene Symbol: SUPT6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320755.1 | NP_001307684.1 | 37, 38 | 5279 | ||
NM_003170.4 | NP_003161.2 | 36, 37 | 5177 | |||
XM_017024978.1 | XP_016880467.1 | 38, 39 | 5338 | |||
XM_017024979.1 | XP_016880468.1 | 36, 37 | 5415 | |||
XM_017024980.1 | XP_016880469.1 | 35, 36 | 5109 | |||
GenBank mRNA | AF070532.1 | - | 5, 6 | 626 | ||
AL833607.1 | - | 37, 38 | 5280 | |||
BC003692.1 | - | 8, 9 | 1169 | |||
BC003696.1 | - | 21, 22 | 3052 | |||
BC033074.1 | - | 13, 14 | 1705 | |||
BC073963.1 | - | 9, 10 | 1476 | |||
BC136522.1 | - | 36, 37 | 5089 | 9020512 | ||
BC136524.1 | - | 36, 37 | 5089 | 9020514 | ||
BC150268.1 | - | 36, 37 | 5091 | 9007230 | ||
D79984.1 | - | 36, 37 | 5091 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009297, U40375, BC072657, AK129069, AK158846, and others.
Gene Ontology Categories:
