siRNA Details
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033549.4 | NP_291027.3 | 1 | 1401 | ||
NM_201627.2 | NP_963921.1 | 1 | 1401 | |||
NR_045218.1 | 1 | 1401 | ||||
XM_006714929.2 | XP_006714992.1 | 1 | 1399 | |||
XM_006714930.2 | XP_006714993.1 | 1 | 1399 | |||
XM_006714931.3 | XP_006714994.1 | 1 | 1399 | |||
XM_006714932.3 | XP_006714995.1 | 1 | 1399 | |||
GenBank mRNA | AB063180.1 | - | 1 | 427 | ||
AB100366.1 | - | 1 | 664 | |||
AB100367.1 | - | 1 | 664 | |||
AK027601.1 | - | 1 | 1374 | |||
AK074052.1 | - | 1 | 498 | |||
AL832960.1 | - | 1 | 278 | |||
AX775789.1 | - | 1 | 140 | |||
AX775791.1 | - | 1 | 140 | |||
AX775793.1 | - | 1 | 1374 | |||
AX775797.1 | - | 1 | 432 | |||
AX775799.1 | - | 1 | 1374 | |||
AX775801.1 | - | 1 | 1374 | |||
AX775803.1 | - | 1 | 432 | |||
BC004956.1 | - | 1 | 606 | |||
BC018765.1 | - | 1 | 432 | |||
BC071887.1 | - | 1 | 606 | |||
BC090953.1 | - | 1 | 944 | 6017628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
