siRNA Details
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031733.2 | NP_001026903.2 | 4 | 1300 | ||
NM_001286694.1 | NP_001273623.1 | 4 | 1410 | |||
NM_001286695.1 | NP_001273624.1 | 3 | 1269 | |||
NM_033429.2 | NP_219501.2 | 5 | 1441 | |||
NR_104583.1 | 2 | 1080 | ||||
GenBank mRNA | AF308287.1 | - | 5 | 889 | ||
AK092628.1 | - | 4 | 522 | |||
BC009516.2 | - | 4 | 617 | |||
BX640702.1 | - | 3 | 458 | |||
BX640817.1 | - | 2 | 1081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
