siRNA Details
Gene Symbol: DEPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114120.2 | NP_001107592.1 | 11 | 2430 | ||
NM_017779.5 | NP_060249.2 | 10 | 1578 | |||
GenBank mRNA | AB281187.1 | - | 11 | 2286 | ||
AB281274.1 | - | 10 | 1434 | |||
AJ278112.1 | - | 10 | 1434 | |||
AK000361.1 | - | 11 | 2296 | |||
AK000490.1 | - | 4 | 755 | |||
AK022792.1 | - | 3 | 1071 | |||
AK225735.1 | - | 11 | 2276 | |||
AK299324.1 | - | 11 | 2291 | |||
AL834208.1 | - | 12 | 1350 | |||
BC065304.1 | - | 10 | 1381 | 6156084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
