siRNA Details
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320559.1 | NP_001307488.1 | 5 | 1752 | ||
NM_001320560.1 | NP_001307489.1 | 5 | 1553 | |||
NM_033027.3 | NP_149016.2 | 5 | 1621 | |||
XM_017007048.1 | XP_016862537.1 | 5 | 4717 | |||
XM_017007049.1 | XP_016862538.1 | 3 | 1005 | |||
GenBank mRNA | AB053121.1 | - | 5 | 1598 | ||
AB063301.1 | - | 5 | 1513 | |||
BC028213.1 | - | 3 | 381 | |||
BC038949.1 | - | 5 | 1568 | 5547938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
