siRNA Details
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040456.1 | NP_001035546.1 | 1 | 238 | ||
NM_001040457.1 | NP_001035547.1 | 1 | 238 | |||
XM_005250511.4 | XP_005250568.1 | 1 | 71 | |||
GenBank mRNA | AF226732.1 | - | 1 | 289 | ||
AK026590.1 | - | 1 | 151 | |||
BC069017.1 | - | 1 | 189 | 3502576 | ||
BG707286.1 | - | 1 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
