siRNA Details
Gene Symbol: ANKDD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182703.5 | NP_874362.3 | 7, 8 | 700 | ||
GenBank mRNA | AK075298.1 | - | 5, 6 | 451 | ||
AK098736.1 | - | 9, 10 | 956 | |||
AK307698.1 | - | 7, 8 | 616 | |||
BC027350.1 | - | 3, 4 | 254 | |||
BC101273.1 | - | 4, 5 | 398 | 40024165 | ||
BC101274.1 | - | 4, 5 | 398 | 40024168 | ||
BC101275.1 | - | 4, 5 | 398 | 40024170 | ||
BC101276.1 | - | 4, 5 | 391 | |||
BC110610.1 | - | 7, 8 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
