siRNA Details
Gene Symbol: MON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014940.3 | NP_055755.1 | 3 | 553 | ||
GenBank mRNA | AB020679.1 | - | 3 | 237 | ||
AF442486.1 | - | 2 | 217 | |||
AK023374.1 | - | 3 | 503 | |||
BC024277.1 | - | 3 | 533 | 4843859 | ||
CF125013.1 | - | 3 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
