siRNA Details
Gene Symbol: MON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014940.3 | NP_055755.1 | 3 | 615 | ||
GenBank mRNA | AB020679.1 | - | 3 | 299 | ||
AF442486.1 | - | 2 | 279 | |||
AK023374.1 | - | 3 | 565 | |||
AK023896.1 | - | 3 | 343 | |||
BC024277.1 | - | 3 | 595 | 4843859 | ||
CF125013.1 | - | 3 | 572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
