siRNA Details
Gene Symbol: MON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014940.3 | NP_055755.1 | 2, 3 | 502 | ||
GenBank mRNA | AB020679.1 | - | 2, 3 | 186 | ||
AF442486.1 | - | 1, 2 | 166 | |||
AK023374.1 | - | 2, 3 | 452 | |||
AK023896.1 | - | 2, 3 | 230 | |||
BC024277.1 | - | 2, 3 | 482 | 4843859 | ||
CF125013.1 | - | 2, 3 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
