siRNA Details
Gene Symbol: C15orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015492.4 | NP_056307.2 | 3 | 3668 | ||
GenBank mRNA | AF063593.1 | - | 2 | 847 | ||
AF143881.1 | - | 1 | 156 | |||
AF495726.1 | - | 3 | 3653 | |||
AK056589.1 | - | 2 | 2234 | |||
AK057025.1 | - | 2 | 1595 | |||
AK125111.1 | - | 3 | 3666 | |||
AK125604.1 | - | 3 | 3637 | |||
AK128205.1 | - | 3 | 3758 | |||
AL117540.1 | - | 2 | 727 | |||
AL360143.1 | - | 2 | 1946 | |||
AL834382.1 | - | 2 | 1814 | |||
BC000540.2 | - | 2 | 1858 | |||
BC001762.1 | - | 2 | 907 | |||
BC006207.2 | - | 2 | 2420 | |||
BC011379.1 | - | 3 | 3640 | 3866082 | ||
BC011905.2 | - | 2 | 816 | |||
BC051235.1 | - | 2 | 919 | |||
BC110367.1 | - | 3 | 3655 | 5494248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
