siRNA Details
Gene Symbol: C15orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015492.4 | NP_056307.2 | 3 | 3313 | ||
GenBank mRNA | AF063593.1 | - | 2 | 492 | ||
AF495726.1 | - | 3 | 3298 | |||
AK056589.1 | - | 2 | 1879 | |||
AK057025.1 | - | 2 | 1240 | |||
AK125111.1 | - | 3 | 3311 | |||
AK125604.1 | - | 3 | 3282 | |||
AK128205.1 | - | 3 | 3403 | |||
AK304663.1 | - | 3 | 1764 | |||
AL117540.1 | - | 2 | 372 | |||
AL360143.1 | - | 2 | 1597 | |||
AL834382.1 | - | 2 | 1459 | |||
BC000540.2 | - | 2 | 1503 | |||
BC001762.1 | - | 2 | 552 | |||
BC006207.2 | - | 2 | 2065 | |||
BC011379.1 | - | 3 | 3285 | 3866082 | ||
BC011905.2 | - | 2 | 461 | |||
BC051235.1 | - | 2 | 564 | |||
BC110367.1 | - | 3 | 3300 | 5494248 | ||
CR533482.1 | - | 2 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
