siRNA Details
Gene Symbol: TTLL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139442.1 | NP_001132914.1 | 4 | 1338 | ||
NM_194252.2 | NP_919228.2 | 4 | 1338 | |||
XM_005251728.2 | XP_005251785.1 | 3 | 639 | |||
XM_005251729.4 | XP_005251786.1 | 4 | 1716 | |||
XM_006716966.3 | XP_006717029.1 | 4 | 1015 | |||
XM_017014309.1 | XP_016869798.1 | 4 | 1015 | |||
GenBank mRNA | AF521886.1 | - | 4 | 1338 | ||
AF521887.1 | - | 3 | 364 | |||
AK125178.1 | - | 3 | 882 | |||
BC171751.1 | - | 2 | 272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
