siRNA Details
Gene Symbol: DOK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 1116 | ||
NM_001256896.1 | NP_001243825.1 | 4 | 825 | |||
NM_001301071.1 | NP_001288000.1 | 7 | 1127 | |||
NM_173660.4 | NP_775931.3 | 7 | 1127 | |||
XM_011513435.2 | XP_011511737.1 | 7 | 1127 | |||
XM_011513437.2 | XP_011511739.1 | 7 | 1116 | |||
XM_017008038.1 | XP_016863527.1 | 5 | 690 | |||
GenBank mRNA | AB220918.1 | - | 7 | 1057 | ||
AK075037.1 | - | 1 | 286 | |||
AK091037.1 | - | 7 | 1111 | |||
AK096456.1 | - | 4 | 825 | |||
BC043568.1 | - | 3 | 464 | |||
BC062369.1 | - | 3 | 519 | |||
BC131544.1 | - | 7 | 1064 | 40085498 | ||
BC141852.1 | - | 7 | 1098 | 8860352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
