siRNA Details
Gene Symbol: FAM219B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321920.1 | NP_001308849.1 | 1 | 177 | ||
NM_001321921.1 | NP_001308850.1 | 1 | 177 | |||
NM_001321922.1 | NP_001308851.1 | 1 | 177 | |||
NM_001321923.1 | NP_001308852.1 | 1 | 177 | |||
NM_020447.4 | NP_065180.1 | 1 | 177 | |||
NR_135841.1 | 1 | 177 | ||||
NR_135842.1 | 1 | 177 | ||||
NR_135843.1 | 1 | 177 | ||||
NR_135844.1 | 1 | 177 | ||||
NR_135845.1 | 1 | 177 | ||||
NR_135846.1 | 1 | 177 | ||||
NR_135847.1 | 1 | 177 | ||||
NR_135848.1 | 1 | 177 | ||||
NR_135849.1 | 1 | 177 | ||||
NR_135850.1 | 1 | 177 | ||||
NR_135851.1 | 1 | 177 | ||||
NR_135852.1 | 1 | 177 | ||||
NR_135853.1 | 1 | 177 | ||||
NR_135854.1 | 1 | 177 | ||||
XM_017022433.1 | XP_016877922.1 | 1 | 499 | |||
GenBank mRNA | AK000005.1 | - | 1 | 79 | ||
AK001572.1 | - | 1 | 166 | |||
AK291020.1 | - | 1 | 164 | |||
AK296403.1 | - | 1 | 164 | |||
BC015637.2 | - | 1 | 121 | 4636850 | ||
DB153105.1 | - | 1 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
