siRNA Details
Gene Symbol: SPEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199339.2 | NP_955371.2 | 3 | 489 | ||
GenBank mRNA | AK097400.1 | - | 1 | 816 | ||
BC033882.1 | - | 3 | 489 | 5742860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
