siRNA Details
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 1, 2 | 374 | ||
XM_005249652.3 | XP_005249709.1 | 1, 2 | 317 | |||
GenBank mRNA | AK096533.1 | - | 1, 2 | 317 | ||
AK294982.1 | - | 1, 2 | 104 | |||
AL834391.1 | - | 1, 2 | 308 | |||
AY054121.1 | - | 1, 2 | 170 | |||
BC062423.1 | - | 1, 2 | 125 | |||
BI560774.1 | - | 1, 2 | 378 | |||
DT217816.1 | - | 1, 2 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
