siRNA Details
Gene Symbol: HOXB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002145.3 | NP_002136.1 | 2 | 562 | ||
XM_005257275.4 | XP_005257332.1 | 2 | 131 | |||
GenBank mRNA | BC074805.2 | - | 2 | 462 | 30915528 | |
BC074806.2 | - | 2 | 462 | 30915314 | ||
M73999.1 | - | 2 | 186 | |||
X14571.1 | - | 1 | 16 | |||
X16665.1 | - | 2 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
