siRNA Details
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 4 | 3303 | ||
NM_001291447.1 | NP_001278376.1 | 4 | 3283 | |||
NM_005407.2 | NP_005398.2 | 2 | 4296 | |||
NR_111966.1 | 3 | 1204 | ||||
NR_111967.1 | 3 | 1184 | ||||
XM_011537064.1 | XP_011535366.1 | 3 | 3634 | |||
XM_011537065.1 | XP_011535367.1 | 3 | 3780 | |||
GenBank mRNA | AB002358.2 | - | 2 | 4283 | ||
AK001276.1 | - | 3 | 922 | |||
AK289358.1 | - | 3 | 1020 | |||
AK293574.1 | - | 4 | 2989 | |||
AK296414.1 | - | 4 | 3102 | |||
BC024245.2 | - | 3 | 997 | |||
BC090958.1 | - | 3 | 1004 | |||
BC136528.1 | - | 2 | 4031 | 9020518 | ||
BC136529.1 | - | 2 | 4031 | 9020519 | ||
X98834.1 | - | 2 | 4183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
