siRNA Details
Gene Symbol: FAM120C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300788.1 | NP_001287717.1 | 1 | 112 | ||
NM_017848.5 | NP_060318.4 | 1 | 89 | |||
NM_198456.2 | NP_940858.2 | 1 | 112 | |||
XM_006724589.3 | XP_006724652.1 | 1 | 85 | |||
GenBank mRNA | AK000513.1 | - | 1 | 56 | ||
AY121803.1 | - | 1 | 112 | |||
AY150025.1 | - | 1 | 89 | |||
BC016138.1 | - | 1 | 112 | |||
BC136413.1 | - | 1 | 86 | 9020401 | ||
BC144081.1 | - | 1 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
