siRNA Details
Gene Symbol: FRMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244959.1 | NP_001231888.1 | 14 | 1625 | ||
NM_001244960.1 | NP_001231889.1 | 14 | 1401 | |||
NM_001244961.1 | NP_001231890.1 | 9 | 937 | |||
NM_001244962.1 | NP_001231891.1 | 2 | 504 | |||
NM_174938.5 | NP_777598.3 | 14 | 1625 | |||
XM_017014588.1 | XP_016870077.1 | 14 | 1315 | |||
XM_017014589.1 | XP_016870078.1 | 9 | 3752 | |||
XM_017014590.1 | XP_016870079.1 | 13 | 1538 | |||
XM_017014591.1 | XP_016870080.1 | 2 | 516 | |||
GenBank mRNA | AK094281.1 | - | 14 | 1326 | ||
AK223597.1 | - | 14 | 1552 | |||
AK297722.1 | - | 14 | 1543 | |||
AY137774.1 | - | 14 | 1409 | |||
BC023560.1 | - | 2 | 504 | |||
BC037253.1 | - | 9 | 937 | 5166212 | ||
BX647549.1 | - | 1 | 187 | |||
EF560742.1 | - | 14 | 1476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
